Assay Details
Target Gene Details
Entrez Gene ID: | 22941 |
Gene Name: | SH3 and multiple ankyrin repeat domains 2 |
Gene Aliases: |
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3 |
Location: |
Chr.11:70467856-71252724 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2 | NM_012309.4 | 25 | 10565 | NP_036441.2 |
| NM_133266.4 | 11 | 8762 | NP_573573.2 | |
| NR_110766.1 | 9 | 6307 | ||
| AA464273.1 | 1 | 111 |
More Information
Set Membership: |
|

Genomic Map