Assay Details
Target Gene Details
Entrez Gene ID: | 27032 |
Gene Name: | ATPase secretory pathway Ca2+ transporting 1 |
Gene Aliases: |
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1 |
Location: |
Chr.3:130850500-131016712 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2C1 | NM_001001485.2 | 2 | 455 | NP_001001485.1 |
| NM_001001486.1 | 1 | 132 | NP_001001486.1 | |
| NM_001001487.1 | 1 | 132 | NP_001001487.1 | |
| NM_001199179.1 | NP_001186108.1 | |||
| NM_001199180.1 | NP_001186109.1 | |||
| NM_001199181.1 | NP_001186110.1 | |||
| NM_001199182.1 | NP_001186111.1 | |||
| NM_001199183.1 | NP_001186112.1 | |||
| NM_001199184.1 | NP_001186113.1 | |||
| NM_001199185.1 | 1 | 274 | NP_001186114.1 | |
| NM_014382.3 | 1 | 274 | NP_055197.2 | |
| XM_005247354.2 | XP_005247411.1 | |||
| XM_005247355.2 | 2 | 502 | XP_005247412.1 | |
| XM_005247356.2 | XP_005247413.1 | |||
| XM_005247358.2 | XP_005247415.1 | |||
| XM_017006164.1 | XP_016861653.1 | |||
| AB037768.1 | BAA92585.1 | |||
| AF181120.1 | 1 | 187 | AAF26295.1 | |
| AF181121.1 | 1 | 187 | AAF26296.1 | |
| AF189723.2 | AAF27813.2 | |||
| AF225981.1 | AAF35375.1 | |||
| AK296470.1 | ||||
| AK299945.1 | ||||
| AK304175.1 | ||||
| AK304374.1 | ||||
| AK314342.1 | 1 | 136 | ||
| AY268374.1 | 1 | 81 | AAP30008.1 | |
| AY268375.1 | 1 | 81 | AAP30009.1 | |
| BC028139.1 | 1 | 144 | AAH28139.1 | |
| DA405579.1 | ||||
| DC343471.1 | 1 | 274 | ||
| DC345627.1 | ||||
| DC422788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005160 | Chr.3:130723453 - 131010147 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map