Assay Details
Target Gene Details
Entrez Gene ID: | 9416 |
Gene Name: | DEAD-box helicase 23 |
Gene Aliases: |
PRPF28, SNRNP100, U5-100K, U5-100KD, prp28 |
Location: |
Chr.12:48829756-48852174 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX23 | NM_004818.2 | NP_004809.2 | ||
| AF026402.1 | AAB87902.1 | |||
| AK128428.1 | ||||
| AK292921.1 | ||||
| AK312379.1 | ||||
| BC002366.2 | AAH02366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826363 | Chr.12:48783444 - 48857206 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75988223] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map