Assay Details
Target Gene Details
Entrez Gene ID: | 678 |
Gene Name: | ZFP36 ring finger protein like 2 |
Gene Aliases: |
BRF2, ERF-2, ERF2, RNF162C, TIS11D |
Location: |
Chr.2:43222402-43226606 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP36L2 | NM_006887.4 | 2 | 1123 | NP_008818.3 |
| BC005010.1 | 2 | 912 | AAH05010.1 | |
| X78992.1 | 2 | 898 | CAA55592.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv581649 | Chr.2:43224732 - 43226264 on Build GRCh38 | Loss |
|
| nsv1007740 | Chr.2:42961811 - 44117003 on Build GRCh38 | Gain |
|
| esv25646 | Chr.2:43224286 - 43227776 on Build GRCh38 | Loss |
|
| nsv581647 | Chr.2:43222999 - 43227061 on Build GRCh38 | Loss |
|
| nsv829097 | Chr.2:43224160 - 43227970 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map