Assay Details
Target Gene Details
Entrez Gene ID: | 138046 |
Gene Name: | RALY RNA binding protein-like |
Gene Aliases: |
HNRPCL3 |
Location: |
Chr.8:84182629-84921844 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALYL | NM_001100392.1 | NP_001093862.1 | ||
| NM_001100393.1 | NP_001093863.1 | |||
| NM_001287243.1 | 1 | 680 | NP_001274172.1 | |
| NM_173848.5 | 1 | 680 | NP_776247.3 | |
| XM_011517462.2 | 1 | 752 | XP_011515764.1 | |
| XM_017013059.1 | XP_016868548.1 | |||
| XM_017013062.1 | XP_016868551.1 | |||
| XM_017013063.1 | XP_016868552.1 | |||
| XM_017013065.1 | XP_016868554.1 | |||
| XM_017013066.1 | XP_016868555.1 | |||
| XM_017013067.1 | 1 | 752 | XP_016868556.1 | |
| XM_017013068.1 | XP_016868557.1 | |||
| XM_017013069.1 | XP_016868558.1 | |||
| XM_017013070.1 | XP_016868559.1 | |||
| XM_017013071.1 | XP_016868560.1 | |||
| XM_017013074.1 | XP_016868563.1 | |||
| XM_017013077.1 | 1 | 919 | XP_016868566.1 | |
| XM_017013078.1 | XP_016868567.1 | |||
| XM_017013079.1 | XP_016868568.1 | |||
| XM_017013082.1 | XP_016868571.1 | |||
| XM_017013085.1 | XP_016868574.1 | |||
| AF495529.1 | 1 | 681 | AAO85516.1 | |
| AF495530.1 | AAO85517.1 | |||
| AK094543.1 | ||||
| AK095026.1 | 1 | 497 | ||
| AK095560.1 | ||||
| DA143105.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611646 | Chr.8:84136455 - 84200142 on Build GRCh38 | Loss |
|
| esv2752268 | Chr.8:83893110 - 84989630 on Build GRCh38 | Gain |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map