Assay Details
Target Gene Details
Entrez Gene ID: | 157567 |
Gene Name: | ankyrin repeat domain 46 |
Gene Aliases: |
ANK-S, GENX-115279 |
Location: |
Chr.8:100509752-100559786 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD46 | NM_001270377.1 | 5 | 1172 | NP_001257306.1 |
| NM_001270378.1 | 5 | 1770 | NP_001257307.1 | |
| NM_001270379.1 | NP_001257308.1 | |||
| NM_198401.3 | 6 | 1277 | NP_940683.1 | |
| XM_006716509.1 | 5 | 1313 | XP_006716572.1 | |
| XM_017013106.1 | 5 | 1235 | XP_016868595.1 | |
| AB077205.1 | 6 | 1275 | BAD23947.1 | |
| AK095235.1 | 5 | 1147 | ||
| AL832842.1 | 5 | 1132 | CAI46169.1 | |
| BC050396.1 | AAH50396.1 | |||
| BC060843.1 | 5 | 1145 | AAH60843.1 | |
| U79297.1 | 1 | 110 |

Genomic Map