Assay Details
Target Gene Details
Entrez Gene ID: | 768 |
Gene Name: | carbonic anhydrase 9 |
Gene Aliases: |
CAIX, MN |
Location: |
Chr.9:35673859-35681159 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA9 | NM_001216.2 | 11 | 1451 | NP_001207.2 |
| AI023541.1 | 1 | 111 | ||
| AJ010588.1 | 11 | 1418 | CAB82444.1 | |
| BC014950.1 | 11 | 1513 | AAH14950.1 | |
| EF122496.1 | 9 | 1273 | ||
| KU177952.1 | ||||
| KU177953.1 | ||||
| X66839.1 | 11 | 1451 | CAA47315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6528 | Chr.9:35666752 - 35700672 on Build GRCh38 | Insertion |
|
| nsv466385 | Chr.9:35662254 - 35705089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map