Assay Details
Target Gene Details
Entrez Gene ID: | 151556 |
Gene Name: | G protein-coupled receptor 155 |
Gene Aliases: |
DEP.7, DEPDC3, PGR22 |
Location: |
Chr.2:174431571-174494001 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR155 | NM_001033045.3 | 17 | 7085 | NP_001028217.1 |
| NM_001267050.1 | 17 | 6982 | NP_001253979.1 | |
| NM_001267051.1 | 15 | 6852 | NP_001253980.1 | |
| NM_152529.6 | 16 | 6936 | NP_689742.4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583667 | Chr.2:174410951 - 174466210 on Build GRCh38 | Loss |
|
| nsv526187 | Chr.2:174421143 - 174474776 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map