Assay Details
Target Gene Details
Entrez Gene ID: | 163786 |
Gene Name: | SAS-6 centriolar assembly protein |
Gene Aliases: |
MCPH14, SAS-6, SAS6 |
Location: |
Chr.1:100082632-100133095 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SASS6 | NM_001304829.1 | 16 | 2133 | NP_001291758.1 |
| NM_194292.2 | 17 | 2213 | NP_919268.1 | |
| XM_017000486.1 | 17 | 1979 | XP_016855975.1 | |
| AK025750.1 | 5 | 511 | ||
| AK302513.1 | 18 | 1963 | ||
| AK307824.1 | 16 | 1999 | ||
| AL834265.1 | 13 | 1476 | CAD38940.1 | |
| AY359522.1 | 17 | 2073 | AAQ57128.1 | |
| BC101026.3 | 16 | 1926 | AAI01027.1 | |
| BX641109.1 | 16 | 2124 | ||
| BX648406.1 | 17 | 1970 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map