Assay Details
Target Gene Details
Entrez Gene ID: | 9547 |
Gene Name: | C-X-C motif chemokine ligand 14 |
Gene Aliases: |
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14 |
Location: |
Chr.5:135570679-135579279 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL14 | NM_004887.4 | 1 | 68 | NP_004878.2 |
| AI743431.1 | 1 | 454 | ||
| AW614568.1 | 1 | 424 |
Target Gene Details
Entrez Gene ID: | 153328 |
Gene Name: | solute carrier family 25 member 48 |
Gene Aliases: |
- |
Location: |
Chr.5:135579002-135888637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A48 | XM_017009090.1 | 1 | 211 | XP_016864579.1 |
| XM_017009092.1 | 1 | 211 | XP_016864581.1 |
More Information
Set Membership: |
|

Genomic Map