Assay Details
Target Gene Details
Entrez Gene ID: | 83990 |
Gene Name: | BRCA1 interacting protein C-terminal helicase 1 |
Gene Aliases: |
BACH1, FANCJ, OF |
Location: |
Chr.17:61679186-61864120 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRIP1 | NM_032043.2 | 20 | 3929 | NP_114432.2 |
| XM_011525332.2 | 21 | 4420 | XP_011523634.1 | |
| XM_011525333.2 | 21 | 4417 | XP_011523635.1 | |
| XM_011525334.2 | 21 | 5077 | XP_011523636.1 | |
| XM_011525335.2 | 21 | 4360 | XP_011523637.1 | |
| XM_011525336.2 | 20 | 4298 | XP_011523638.1 | |
| XM_011525337.2 | 19 | 4217 | XP_011523639.1 | |
| XM_011525338.2 | 17 | 3297 | XP_011523640.1 | |
| XM_017025200.1 | 16 | 3237 | XP_016880689.1 | |
| XM_017025201.1 | 16 | 4002 | XP_016880690.1 | |
| XM_017025202.1 | 8 | 2365 | XP_016880691.1 | |
| XM_017025203.1 | 9 | 1877 | XP_016880692.1 | |
| AF360549.1 | 20 | 3764 | AAK38111.1 | |
| BC101472.1 | 20 | 3679 | AAI01473.1 | |
| BC101474.1 | 20 | 3679 | AAI01475.1 |
More Information
Set Membership: |
|

Genomic Map