Assay Details
Target Gene Details
Entrez Gene ID: | 51361 |
Gene Name: | hook microtubule-tethering protein 1 |
Gene Aliases: |
HK1 |
Location: |
Chr.1:59814861-59876378 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOOK1 | NM_015888.4 | NP_056972.1 | ||
| XM_006710676.1 | XP_006710739.1 | |||
| XM_011541562.2 | XP_011539864.1 | |||
| XM_011541563.1 | XP_011539865.1 | |||
| XM_017001424.1 | XP_016856913.1 | |||
| AF044923.1 | AAC09298.1 | |||
| AK097735.1 | ||||
| AK292314.1 | ||||
| AK301768.1 | ||||
| BC011621.2 | AAH11621.1 | |||
| BC107857.1 | AAI07858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012703 | Chr.1:59796670 - 59897484 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map