Assay Details
Target Gene Details
Entrez Gene ID: | 63036 |
Gene Name: | chymotrypsin like elastase family member 2A |
Gene Aliases: |
ELA2A, PE-1 |
Location: |
Chr.1:15456728-15472091 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELA2A | NM_033440.2 | 7 | 742 | NP_254275.1 |
| AK312198.1 | 7 | 738 | ||
| BC007031.1 | 7 | 741 | AAH07031.1 | |
| BC069331.1 | 7 | 741 | AAH69331.1 | |
| BC069432.1 | 7 | 741 | AAH69432.1 | |
| BQ787677.1 | 2 | 193 | ||
| CR450291.1 | 7 | 716 | CAG29287.1 | |
| D00236.1 | 7 | 720 | BAA00165.1 | |
| M16631.1 | 7 | 737 | AAA52374.1 | |
| M16652.1 | 7 | 742 | AAA52380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161190 | Chr.1:15463238 - 15484851 on Build GRCh38 | Duplication |
|
| esv2744308 | Chr.1:15455456 - 15475909 on Build GRCh38 | Deletion |
|
| nsv441683 | Chr.1:15465929 - 15484726 on Build GRCh38 | Loss |
|
| esv3548899 | Chr.1:15454506 - 15475973 on Build GRCh38 | Deletion |
|
| nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss |
|
| nsv1114929 | Chr.1:15455856 - 15475193 on Build GRCh38 | Deletion |
|
| esv2761582 | Chr.1:15457276 - 15475000 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map