Assay Details
Target Gene Details
Entrez Gene ID: | 1259 |
Gene Name: | cyclic nucleotide gated channel alpha 1 |
Gene Aliases: |
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49 |
Location: |
Chr.4:47935015-48016718 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNGA1 | NM_000087.3 | 11 | 1764 | NP_000078.2 |
| NM_001142564.1 | 10 | 1846 | NP_001136036.1 | |
| XM_005248049.4 | 9 | 1736 | XP_005248106.2 | |
| XM_011513623.2 | 10 | 1827 | XP_011511925.1 | |
| XM_017007712.1 | 11 | 1819 | XP_016863201.1 | |
| AK292021.1 | 11 | 1764 | ||
| M84741.1 | 9 | 1522 | AAA52010.1 | |
| S42457.1 | 10 | 1861 | AAB22778.1 |
Target Gene Details
Entrez Gene ID: | 101927157 |
Gene Name: | uncharacterized LOC101927157 |
Gene Aliases: |
- |
Location: |
Chr.4:47914227-47990737 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927157 | NR_125879.1 |
More Information
Set Membership: |
|

Genomic Map