Assay Details
Target Gene Details
Entrez Gene ID: | 56884 |
Gene Name: | follistatin like 5 |
Gene Aliases: |
- |
Location: |
Chr.4:161383892-162164034 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSTL5 | NM_001128427.2 | 16 | 3999 | NP_001121899.1 |
| NM_001128428.2 | 15 | 3972 | NP_001121900.1 | |
| NM_020116.4 | 16 | 4002 | NP_064501.2 | |
| XM_011532126.1 | 15 | 3941 | XP_011530428.1 | |
| AB033089.1 | 16 | 3929 | ||
| AL137695.1 | 13 | 3345 | CAB70877.1 | |
| BC036502.2 | 16 | 4003 | AAH36502.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830133 | Chr.4:161325036 - 161458867 on Build GRCh38 | Gain |
|
| nsv1031902 | Chr.4:161384208 - 161830698 on Build GRCh38 | Gain |
|
| dgv9344n54 | Chr.4:160658267 - 161986681 on Build GRCh38 | Gain |
|
| dgv5454n100 | Chr.4:161000260 - 162613846 on Build GRCh38 | Gain |
|
| esv3602905 | Chr.4:161346340 - 161396269 on Build GRCh38 | Gain |
|
| nsv1032175 | Chr.4:160266174 - 162269517 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map