Assay Details
Target Gene Details
Entrez Gene ID: | 2949 |
Gene Name: | glutathione S-transferase mu 5 |
Gene Aliases: |
GSTM5-5, GTM5 |
Location: |
Chr.1:109711751-109718268 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTM5 | NM_000851.3 | NP_000842.2 | ||
| XM_005270784.4 | XP_005270841.1 | |||
| XM_005270785.4 | XP_005270842.1 | |||
| AK125820.1 | 2 | 3691 | ||
| AK127250.1 | 5 | 3431 | BAC86900.1 | |
| AK289673.1 | ||||
| AK315260.1 | ||||
| BC058881.1 | AAH58881.1 | |||
| BI824480.1 | ||||
| L02321.1 | AAA20040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001758 | Chr.1:109650144 - 109721978 on Build GRCh38 | Gain |
|
| nsv1007399 | Chr.1:109691597 - 109726206 on Build GRCh38 | Loss |
|
| nsv1003914 | Chr.1:109684109 - 109741677 on Build GRCh38 | Gain |
|
| dgv270n100 | Chr.1:109681750 - 109727220 on Build GRCh38 | Gain |
|
| esv1003982 | Chr.1:109672531 - 109717528 on Build GRCh38 | Deletion |
|
| nsv2310 | Chr.1:109674297 - 109718036 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map