Assay Details
Target Gene Details
Entrez Gene ID: | 51527 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
C14orf129, HSPC210 |
Location: |
Chr.14:96363452-96387290 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSKIP | NM_001271904.1 | 4 | 1366 | NP_001258833.1 |
| NM_001271905.1 | 4 | 919 | NP_001258834.1 | |
| NM_001271906.1 | 3 | 851 | NP_001258835.1 | |
| NM_016472.4 | 4 | 952 | NP_057556.2 | |
| XM_017021365.1 | 4 | 1060 | XP_016876854.1 | |
| AF151044.1 | 4 | 870 | AAF36130.1 | |
| AK000796.1 | 4 | 878 | BAA91380.1 | |
| AK094654.1 | 4 | 1368 | ||
| BC004818.2 | 3 | 838 | AAH04818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635423 | Chr.14:96385068 - 96403803 on Build GRCh38 | Gain |
|
| nsv565649 | Chr.14:96377253 - 96453283 on Build GRCh38 | Gain |
|

Genomic Map