Assay Details
Target Gene Details
Entrez Gene ID: | 55601 |
Gene Name: | DEXD/H-box helicase 60 |
Gene Aliases: |
- |
Location: |
Chr.4:168216291-168318807 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX60 | NM_017631.5 | NP_060101.3 | ||
| XM_011532103.2 | XP_011530405.1 | |||
| AK000042.1 | BAA90901.1 | |||
| AK223184.1 | BAD96904.1 | |||
| AK225191.1 | ||||
| BC038115.2 | AAH38115.1 | |||
| BC074781.2 | AAH74781.1 | |||
| BX648655.1 | CAE46197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4596 | Chr.4:168228336 - 168263735 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map