Assay Details
Target Gene Details
Entrez Gene ID: | 871 |
Gene Name: | serpin family H member 1 |
Gene Aliases: |
AsTP3, CBP1, CBP2, HSP47, OI10, PIG14, PPROM, RA-A47, SERPINH2, gp46 |
Location: |
Chr.11:75562056-75572804 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINH1 | NM_001207014.1 | NP_001193943.1 | ||
| NM_001235.3 | NP_001226.2 | |||
| XM_011545327.1 | XP_011543629.1 | |||
| AB044778.1 | BAA96788.1 | |||
| AB044779.1 | BAA96789.1 | |||
| AK075504.1 | ||||
| AK094047.1 | ||||
| AK122936.1 | ||||
| AK130870.1 | ||||
| AK290124.1 | ||||
| AK297811.1 | ||||
| AY264853.1 | AAP93914.1 | |||
| AY744367.1 | AAU95378.1 | |||
| BC014623.2 | AAH14623.1 | |||
| BC036298.1 | AAH36298.2 | |||
| BC070087.1 | AAH70087.1 | |||
| BT007094.1 | AAP35758.1 | |||
| D83174.1 | BAA11829.1 | |||
| X61598.1 | CAA43795.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525193 | Chr.11:75565133 - 75576296 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map