Assay Details
Target Gene Details
Entrez Gene ID: | 5277 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class A |
Gene Aliases: |
GPI3, MCAHS2, PIG-A, PNH1 |
Location: |
Chr.X:15319451-15335554 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGA | NM_002641.3 | 6 | 2439 | NP_002632.1 |
| NM_020473.3 | 5 | 1662 | NP_065206.3 | |
| NR_033835.1 | 6 | 2065 | ||
| NR_033836.1 | 6 | 1781 | ||
| XM_011545539.2 | 5 | 3263 | XP_011543841.1 | |
| XM_017029581.1 | 5 | 2386 | XP_016885070.1 | |
| AK098047.1 | 5 | 1875 | ||
| BC038236.1 | 5 | 2396 | AAH38236.1 | |
| D11466.1 | 6 | 2408 | BAA02019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522729 | Chr.X:15283411 - 15334052 on Build GRCh38 | Gain |
|
| nsv517938 | Chr.X:15296656 - 15331048 on Build GRCh38 | Gain |
|
| nsv528343 | Chr.X:15271931 - 15385229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map