Assay Details
Target Gene Details
Entrez Gene ID: | 1514 |
Gene Name: | cathepsin L |
Gene Aliases: |
CATL, CTSL1, MEP |
Location: |
Chr.9:87725437-87731469 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSL | NM_001257971.1 | 8 | 1338 | NP_001244900.1 |
| NM_001257972.1 | 8 | 1279 | NP_001244901.1 | |
| NM_001257973.1 | 6 | 853 | NP_001244902.1 | |
| NM_001912.4 | 8 | 1428 | NP_001903.1 | |
| NM_145918.2 | 8 | 1283 | NP_666023.1 | |
| XM_005251716.3 | 9 | 2158 | XP_005251773.1 | |
| XM_011518263.1 | 7 | 1850 | XP_011516565.1 | |
| XM_017014293.1 | 7 | 1787 | XP_016869782.1 | |
| AF217997.1 | 6 | 778 | AAG17239.1 | |
| AK055599.1 | 8 | 1966 | ||
| AK075100.1 | 8 | 1234 | ||
| AL832167.1 | 8 | 1226 | ||
| AW270438.1 | 1 | 313 | ||
| BC012612.1 | 8 | 1276 | AAH12612.1 | |
| BC142983.1 | 8 | 1223 | ||
| BM680778.1 | 1 | 303 | ||
| BX537395.1 | 8 | 1219 | CAD97637.1 | |
| BX647101.1 | 8 | 1223 | ||
| BX647102.1 | 8 | 1222 | ||
| BX647413.1 | 8 | 1223 | ||
| BX647434.1 | 8 | 1281 | ||
| BX647435.1 | 8 | 1278 | ||
| BX648848.1 | 8 | 1233 | ||
| BX648849.1 | 8 | 1223 | ||
| BX649140.1 | 8 | 1279 | ||
| CB963086.1 | 1 | 125 | ||
| X12451.1 | 8 | 1366 | CAA30981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759703 | Chr.9:87403812 - 87810065 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map