Assay Details
Target Gene Details
Entrez Gene ID: | 375519 |
Gene Name: | gap junction protein beta 7 |
Gene Aliases: |
CX25, bA136M9.1, connexin25 |
Location: |
Chr.6:87282978-87329278 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB7 | NM_198568.2 | 3 | 1840 | NP_940970.1 |
| XM_005248716.2 | 2 | 1577 | XP_005248773.1 | |
| AK127554.1 | 2 | 1665 | ||
| BC057813.1 | 3 | 1840 | AAH57813.1 | |
| BM976147.1 | 1 | 459 |
More Information
Set Membership: |
|

Genomic Map