Assay Details
Target Gene Details
Entrez Gene ID: | 27350 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 3C |
Gene Aliases: |
A3C, APOBEC1L, ARDC2, ARDC4, ARP5, PBI, bK150C2.3 |
Location: |
Chr.22:39014260-39018838 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC3C | NM_014508.2 | 4 | 801 | NP_055323.2 |
| AB208979.1 | 4 | 766 | BAD92216.1 | |
| AF165520.1 | 4 | 805 | AAF86650.1 | |
| BC011739.2 | 4 | 801 | AAH11739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763703 | Chr.22:38955461 - 39034450 on Build GRCh38 | Loss |
|
| esv2724241 | Chr.22:39014940 - 39028280 on Build GRCh38 | Deletion |
|
| esv3647751 | Chr.22:38997523 - 39050430 on Build GRCh38 | Gain |
|
| esv3647753 | Chr.22:39004677 - 39029340 on Build GRCh38 | Gain |
|
| nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss |
|
| esv1001536 | Chr.22:39018312 - 39018821 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map