Assay Details
Target Gene Details
Entrez Gene ID: | 8484 |
Gene Name: | galanin receptor 3 |
Gene Aliases: |
- |
Location: |
Chr.22:37823382-37825495 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALR3 | NM_003614.1 | 2 | 826 | NP_003605.1 |
| AF067733.1 | 2 | 801 | AAC18860.1 | |
| AF073799.1 | 2 | 826 | AAC35944.1 | |
| AY587582.1 | 2 | 801 | AAS98215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117618411] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map