Assay Details
Target Gene Details
Entrez Gene ID: | 403313 |
Gene Name: | phospholipid phosphatase 6 |
Gene Aliases: |
PDP1, PPAPDC2, PSDP, bA6J24.6 |
Location: |
Chr.9:4662294-4665274 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP6 | NM_203453.3 | 1 | 269 | NP_982278.3 |
| AK074672.1 | 1 | 248 | ||
| AK128369.1 | 1 | 229 | ||
| AK296773.1 | 2 | 226 | ||
| BC038108.2 | 1 | 268 | AAH38108.2 | |
| DA416947.1 | 1 | 269 |
Target Gene Details
Entrez Gene ID: | 55064 |
Gene Name: | spermatogenesis associated 6 like |
Gene Aliases: |
C9orf68, bA6J24.2 |
Location: |
Chr.9:4540092-4666674 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA6L | NM_001039395.3 | NP_001034484.3 | ||
| XM_006716795.2 | XP_006716858.1 | |||
| XM_006716798.3 | XP_006716861.1 | |||
| XM_006716799.3 | XP_006716862.1 | |||
| XM_006716801.2 | XP_006716864.1 | |||
| XM_011517954.1 | XP_011516256.1 | |||
| XM_011517956.1 | XP_011516258.1 | |||
| XM_011517957.1 | XP_011516259.1 | |||
| XM_017014881.1 | XP_016870370.1 | |||
| XM_017014882.1 | XP_016870371.1 | |||
| XM_017014883.1 | XP_016870372.1 | |||
| XM_017014885.1 | XP_016870374.1 | |||
| AK000920.1 | BAA91425.1 | |||
| AK295831.1 | ||||
| AK301790.1 | ||||
| BX649030.1 | ||||
| DA234263.1 |
More Information
Set Membership: |
|

Genomic Map