Assay Details
Target Gene Details
Entrez Gene ID: | 79883 |
Gene Name: | podocan like 1 |
Gene Aliases: |
SLRR5B |
Location: |
Chr.19:13931187-13953392 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PODNL1 | NM_001146254.1 | 8 | 1308 | NP_001139726.1 |
| NM_001146255.1 | 6 | 1041 | NP_001139727.1 | |
| NM_024825.3 | 8 | 1332 | NP_079101.3 | |
| XM_006722901.1 | 8 | 1265 | XP_006722964.1 | |
| XM_006722902.1 | 7 | 1130 | XP_006722965.1 | |
| XM_006722903.1 | 6 | 998 | XP_006722966.1 | |
| XM_011528308.1 | 8 | 1299 | XP_011526610.1 | |
| XM_011528309.2 | 7 | 1224 | XP_011526611.1 | |
| XM_011528310.2 | 7 | 1187 | XP_011526612.1 | |
| XM_011528311.1 | 7 | 1166 | XP_011526613.1 | |
| XM_011528313.1 | 7 | 1234 | XP_011526615.1 | |
| XM_011528314.1 | 7 | 1155 | XP_011526616.1 | |
| XM_011528316.1 | 5 | 967 | XP_011526618.1 | |
| XM_017027313.1 | 7 | 1189 | XP_016882802.1 | |
| AK027100.1 | 6 | 1032 | BAB15657.1 | |
| AK290731.1 | 8 | 1265 | ||
| AK296101.1 | 6 | 1028 | ||
| AK298489.1 | 8 | 1308 | ||
| BC057786.1 | 8 | 1332 | AAH57786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953979 | Chr.19:13795787 - 14006888 on Build GRCh38 | Deletion |
|
| nsv833757 | Chr.19:13725904 - 13951988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map