Assay Details
Target Gene Details
Entrez Gene ID: | 3094 |
Gene Name: | histidine triad nucleotide binding protein 1 |
Gene Aliases: |
HINT, NMAN, PKCI-1, PRKCNH1 |
Location: |
Chr.5:131159283-131165348 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HINT1 | NM_005340.6 | NP_005331.1 | ||
| NR_024610.2 | ||||
| NR_024611.2 | ||||
| NR_073488.1 | ||||
| NR_134494.1 | ||||
| NR_134495.1 | ||||
| XM_017009410.1 | XP_016864899.1 | |||
| AK026557.1 | BAB15500.1 | |||
| BC001287.1 | AAH01287.1 | |||
| BC007090.1 | AAH07090.1 | |||
| BF967059.1 | ||||
| BF970603.1 | ||||
| BG393782.1 | ||||
| BG527380.1 | ||||
| BG708765.1 | ||||
| BG942577.1 | ||||
| CD639104.1 | ||||
| CK002748.1 | ||||
| CN290233.1 | ||||
| CN290239.1 | ||||
| CR457048.1 | CAG33329.1 | |||
| DB125655.1 | ||||
| DB289264.1 | 2 | 329 | ||
| U27143.1 | AAA82926.1 | |||
| U51004.1 | AAC71077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575881 | Chr.5:130637365 - 131735693 on Build GRCh38 | Gain |
|
| dgv5808n100 | Chr.5:130705748 - 131180675 on Build GRCh38 | Gain |
|
| esv2759379 | Chr.5:131103304 - 131256219 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map