Assay Details
Target Gene Details
Entrez Gene ID: | 266629 |
Gene Name: | SEC14 like lipid binding 3 |
Gene Aliases: |
TAP2 |
Location: |
Chr.22:30447959-30472047 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC14L3 | NM_001257378.1 | 14 | 2207 | NP_001244307.1 |
| NM_001257379.1 | 13 | 2037 | NP_001244308.1 | |
| NM_001257382.1 | 13 | 2107 | NP_001244311.1 | |
| NM_174975.4 | 12 | 1989 | NP_777635.1 | |
| XM_011530128.2 | XP_011528430.1 | |||
| AK131358.1 | ||||
| AY240872.1 | 12 | 1989 | AAO52677.1 | |
| BE219833.1 | 1 | 101 | ||
| BQ186227.1 | 1 | 494 | ||
| EL947765.1 | 1 | 381 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588886 | Chr.22:30440326 - 30472431 on Build GRCh38 | Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map