Assay Details
Target Gene Details
Entrez Gene ID: | 7412 |
Gene Name: | vascular cell adhesion molecule 1 |
Gene Aliases: |
CD106, INCAM-100 |
Location: |
Chr.1:100719640-100739045 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCAM1 | NM_001078.3 | 9 | 2380 | NP_001069.1 |
| NM_001199834.1 | 9 | 2194 | NP_001186763.1 | |
| NM_080682.2 | 8 | 2104 | NP_542413.1 | |
| AK223266.1 | 9 | 2279 | BAD96986.1 | |
| AK291732.1 | 9 | 2278 | ||
| AK296692.1 | 9 | 2092 | ||
| BC017276.2 | 9 | 2260 | AAH17276.3 | |
| BC068490.1 | 9 | 2278 | AAH68490.2 | |
| BC085003.1 | 9 | 2260 | AAH85003.1 | |
| BF791565.1 | 1 | 79 | ||
| BF968655.1 | 3 | 403 | ||
| CR749464.1 | 1 | 3050 | CAH18296.1 | |
| M30257.1 | 8 | 1989 | AAA51917.1 | |
| M60335.1 | 9 | 2258 | AAA61269.1 | |
| X53051.1 | 9 | 2159 | CAA37218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
| nsv546890 | Chr.1:100736666 - 100751050 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map