Assay Details
Target Gene Details
Entrez Gene ID: | 1964 |
Gene Name: | eukaryotic translation initiation factor 1A, X-linked |
Gene Aliases: |
EIF1A, EIF1AP1, EIF4C, eIF-1A, eIF-4C |
Location: |
Chr.X:20124518-20141848 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1AX | NM_001412.3 | NP_001403.1 | ||
| BC000793.1 | AAH00793.1 | |||
| BC067851.1 | AAH67851.1 | |||
| L18960.1 | AAA19812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25136 | Chr.X:20125506 - 20128380 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map