Assay Details
Target Gene Details
Entrez Gene ID: | 50804 |
Gene Name: | myelin expression factor 2 |
Gene Aliases: |
HsT18564, MEF-2, MST156, MSTP156, myEF-2 |
Location: |
Chr.15:48134632-48178517 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYEF2 | XM_005254425.4 | 16 | 8087 | XP_005254482.2 |
| XM_017022285.1 | 15 | 8036 | XP_016877774.1 | |
| XM_017022286.1 | 15 | 8015 | XP_016877775.1 | |
| XM_017022287.1 | 14 | 7964 | XP_016877776.1 | |
| XM_017022291.1 | 14 | 7807 | XP_016877780.1 | |
| XM_017022292.1 | 13 | 7756 | XP_016877781.1 |
Target Gene Details
Entrez Gene ID: | 283652 |
Gene Name: | solute carrier family 24 member 5 |
Gene Aliases: |
JSX, NCKX5, OCA6, SHEP4 |
Location: |
Chr.15:48120972-48142392 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC24A5 | NM_205850.2 | 6 | 833 | NP_995322.1 |
| XM_017022079.1 | 5 | 1934 | XP_016877568.1 | |
| XM_017022080.1 | 5 | 2154 | XP_016877569.1 | |
| AF348468.1 | 6 | 833 | AAQ15116.1 | |
| BC113628.1 | 6 | 788 | ||
| BC113630.1 | 6 | 788 | ||
| BC143950.1 | 6 | 788 | ||
| DQ665306.1 | 6 | 781 | ||
| DQ665307.1 | 5 | 601 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832999 | Chr.15:47960097 - 48160208 on Build GRCh38 | Gain |
|
| nsv569338 | Chr.15:48104611 - 48160602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map