Assay Details
Target Gene Details
Entrez Gene ID: | 340578 |
Gene Name: | DDB1 and CUL4 associated factor 12 like 2 |
Gene Aliases: |
WDR40C |
Location: |
Chr.X:126163499-126166097 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF12L2 | NM_001013628.2 | 1 | 838 | NP_001013650.1 |
| BC136673.1 | 2 | 746 | ||
| BC136674.1 | 1 | 838 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524503 | Chr.X:126044730 - 126316838 on Build GRCh38 | Gain |
|
| nsv519795 | Chr.X:126124167 - 126251354 on Build GRCh38 | Gain |
|

Genomic Map