Assay Details
Target Gene Details
Entrez Gene ID: | 121340 |
Gene Name: | Sp7 transcription factor |
Gene Aliases: |
OI11, OI12, OSX, osterix |
Location: |
Chr.12:53326575-53344793 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP7 | NM_001173467.2 | 3 | 1350 | NP_001166938.1 |
| NM_001300837.1 | 3 | 1388 | NP_001287766.1 | |
| NM_152860.1 | 2 | 1172 | NP_690599.1 | |
| XM_011537900.2 | 3 | 1439 | XP_011536202.1 | |
| AF466179.1 | 2 | 1173 | AAO33377.1 | |
| AF477981.1 | 2 | 1172 | AAL84281.1 | |
| AK128520.1 | 3 | 1349 | ||
| AY150673.1 | 2 | 1111 | AAN85556.1 | |
| AY150674.1 | 2 | 1119 | AAN85557.1 | |
| BC101549.1 | 2 | 1098 | AAI01550.1 | |
| BC113613.1 | 2 | 1098 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map