Assay Details
Target Gene Details
Entrez Gene ID: | 1051 |
Gene Name: | CCAAT/enhancer binding protein beta |
Gene Aliases: |
C/EBP-beta, IL6DBP, NF-IL6, TCF5 |
Location: |
Chr.20:50190583-50192690 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPB | NM_001285878.1 | 1 | 1390 | NP_001272807.1 |
| NM_001285879.1 | 1 | 1390 | NP_001272808.1 | |
| NM_005194.3 | 1 | 1390 | NP_005185.2 | |
| AF289608.1 | 1 | 1131 | AAL55792.1 | |
| AK291536.1 | 1 | 1143 | ||
| BC005132.2 | 1 | 317 | AAH05132.2 | |
| BC007538.1 | 1 | 1119 | AAH07538.1 | |
| BC021931.1 | 1 | 1134 | AAH21931.1 | |
| D63161.1 | BAA09637.1 |
Target Gene Details
Entrez Gene ID: | 101927559 |
Gene Name: | CEBPB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:50184603-50192069 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPB-AS1 | NR_125739.1 | 1 | 98 | |
| DA253693.1 | 1 | 98 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv702n67 | Chr.20:50190063 - 50192790 on Build GRCh38 | Gain |
|
| nsv828736 | Chr.20:50189040 - 50193150 on Build GRCh38 | Gain |
|
| nsv1061982 | Chr.20:50111064 - 50483508 on Build GRCh38 | Loss |
|
| esv34129 | Chr.20:50132910 - 50304579 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map