Assay Details
Target Gene Details
Entrez Gene ID: | 80055 |
Gene Name: | post-GPI attachment to proteins 1 |
Gene Aliases: |
Bst1, ISPD3024, MRT42, SPG67 |
Location: |
Chr.2:196833004-196926995 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAP1 | NM_001321099.1 | 28 | 5675 | NP_001308028.1 |
| NM_001321100.1 | 26 | 5535 | NP_001308029.1 | |
| NM_024989.3 | 27 | 5590 | NP_079265.2 | |
| XM_017004992.1 | 27 | 5422 | XP_016860481.1 | |
| XM_017004993.1 | 28 | 5992 | XP_016860482.1 | |
| XM_017004994.1 | 26 | 5366 | XP_016860483.1 | |
| AK001646.1 | 1 | 1032 | ||
| AK124764.1 | 4 | 3187 | ||
| AL050078.1 | 1 | 2087 | ||
| BX648642.1 | 28 | 5599 | CAH10543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575323 | Chr.2:196674745 - 197095661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map