Assay Details
Target Gene Details
Entrez Gene ID: | 10249 |
Gene Name: | glycine-N-acyltransferase |
Gene Aliases: |
ACGNAT, CAT, GAT |
Location: |
Chr.11:58708757-58731974 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLYAT | NM_201648.2 | 6 | 1651 | NP_964011.2 |
| XM_017017087.1 | 6 | 1714 | XP_016872576.1 | |
| AI253073.1 | ||||
| AI793012.1 | ||||
| AI793201.1 | 1 | 408 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832168 | Chr.11:58626587 - 58797178 on Build GRCh38 | Gain |
|
| nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain |
|
| nsv329 | Chr.11:58683571 - 58710817 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map