Assay Details
Target Gene Details
Entrez Gene ID: | 6197 |
Gene Name: | ribosomal protein S6 kinase A3 |
Gene Aliases: |
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2 |
Location: |
Chr.X:20149911-20267514 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA3 | NM_004586.2 | 22 | 6841 | NP_004577.1 |
| XM_005274573.2 | 22 | 6842 | XP_005274630.1 | |
| XM_005274577.2 | 21 | 6778 | XP_005274634.1 | |
| XM_006724507.3 | 24 | 6944 | XP_006724570.1 | |
| XM_011545555.1 | 23 | 2802 | XP_011543857.1 | |
| XM_011545556.1 | 22 | 6860 | XP_011543858.1 | |
| XM_011545557.2 | 24 | 6981 | XP_011543859.1 | |
| XM_011545558.2 | 24 | 7045 | XP_011543860.1 | |
| XM_011545559.1 | 24 | 7042 | XP_011543861.1 | |
| XM_011545560.1 | 23 | 6897 | XP_011543862.1 | |
| XM_011545561.1 | 22 | 6823 | XP_011543863.1 | |
| XM_011545562.1 | 21 | 6799 | XP_011543864.1 | |
| XM_011545563.2 | 24 | 6961 | XP_011543865.1 | |
| XM_017029713.1 | 24 | 6949 | XP_016885202.1 | |
| XM_017029714.1 | 24 | 7025 | XP_016885203.1 | |
| XM_017029715.1 | 24 | 7024 | XP_016885204.1 | |
| XM_017029716.1 | 24 | 7099 | XP_016885205.1 | |
| XM_017029717.1 | 22 | 6805 | XP_016885206.1 | |
| XM_017029718.1 | 21 | 6781 | XP_016885207.1 | |
| XM_017029719.1 | 22 | 6802 | XP_016885208.1 | |
| AK022869.1 | 1 | 1834 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526033 | Chr.X:20149203 - 20206450 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map