Assay Details
Target Gene Details
Entrez Gene ID: | 3248 |
Gene Name: | hydroxyprostaglandin dehydrogenase 15-(NAD) |
Gene Aliases: |
15-PGDH, PGDH, PGDH1, PHOAR1, SDR36C1 |
Location: |
Chr.4:174490177-174522898 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPGD | NM_000860.5 | 7 | 2420 | NP_000851.2 |
| NM_001145816.2 | 6 | 2256 | NP_001139288.1 | |
| NM_001256301.1 | 7 | 2150 | NP_001243230.1 | |
| NM_001256305.1 | 5 | 2179 | NP_001243234.1 | |
| NM_001256306.1 | 5 | 2216 | NP_001243235.1 | |
| NM_001256307.1 | 6 | 2313 | NP_001243236.1 | |
| AK058013.1 | 7 | 2009 | ||
| BC018986.2 | 7 | 1982 | AAH18986.1 | |
| J05594.1 | 7 | 1990 | AAA89174.1 | |
| L76465.1 | 7 | 1990 | AAA89175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525487 | Chr.4:174478953 - 174549716 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map