Assay Details
Target Gene Details
Entrez Gene ID: | 201627 |
Gene Name: | DENN domain containing 6A |
Gene Aliases: |
AFI1A, FAM116A |
Location: |
Chr.3:57625454-57693118 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND6A | NM_152678.2 | 20 | 2555 | NP_689891.1 |
| XM_006713019.3 | 20 | 2581 | XP_006713082.1 | |
| XM_006713020.2 | 21 | 2787 | XP_006713083.1 | |
| XM_017005864.1 | 21 | 2790 | XP_016861353.1 | |
| AK074156.1 | 20 | 2542 | ||
| AK092288.1 | BAC03852.1 | |||
| BC040291.1 | 20 | 2543 | AAH40291.1 | |
| BC054343.1 | 11 | 1638 | AAH54343.1 |
Target Gene Details
Entrez Gene ID: | 201626 |
Gene Name: | phosphodiesterase 12 |
Gene Aliases: |
2'-PDE, 2-PDE |
Location: |
Chr.3:57556247-57656480 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE12 | NM_001322176.1 | NP_001309105.1 | ||
| BM713922.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3596235 | Chr.3:57626005 - 57627619 on Build GRCh38 | Loss |
|
| nsv1008684 | Chr.3:57537986 - 57766490 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map