Assay Details
Target Gene Details
Entrez Gene ID: | 8898 |
Gene Name: | myotubularin related protein 2 |
Gene Aliases: |
CMT4B, CMT4B1 |
Location: |
Chr.11:95832880-95924207 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR2 | NM_001243571.1 | 18 | 4713 | NP_001230500.1 |
| NM_016156.5 | 15 | 4446 | NP_057240.3 | |
| NM_201278.2 | 17 | 4640 | NP_958435.1 | |
| NM_201281.2 | 16 | 4517 | NP_958438.1 | |
| XM_005274374.2 | 16 | 4358 | XP_005274431.1 | |
| XM_005274375.2 | 16 | 4472 | XP_005274432.1 | |
| XM_006718934.2 | 17 | 4552 | XP_006718997.1 | |
| XM_006718935.2 | 17 | 4482 | XP_006718998.1 | |
| XM_006718936.3 | 16 | 4484 | XP_006718999.1 | |
| XM_011543058.2 | 18 | 4574 | XP_011541360.1 | |
| XM_011543059.2 | 18 | 4408 | XP_011541361.1 | |
| XM_017018517.1 | 17 | 4579 | XP_016874006.1 | |
| XM_017018518.1 | 16 | 4361 | XP_016874007.1 | |
| AB028996.1 | 15 | 4534 | ||
| AI360884.1 | 1 | 148 | ||
| DA831307.1 | 1 | 243 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579857 | Chr.11:95832676 - 95833871 on Build GRCh38 | Loss |
|
| esv3627359 | Chr.11:95832628 - 95833966 on Build GRCh38 | Loss |
|
| nsv832235 | Chr.11:95753612 - 95935071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map