Assay Details
Target Gene Details
Entrez Gene ID: | 113402 |
Gene Name: | SFT2 domain containing 1 |
Gene Aliases: |
C6orf83, pRGR1 |
Location: |
Chr.6:166319728-166342606 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFT2D1 | NM_145169.2 | 2 | 241 | NP_660152.1 |
| NR_130112.1 | 3 | 264 | ||
| NR_130113.1 | 3 | 264 | ||
| NR_130114.1 | 3 | 255 | ||
| AF041429.1 | 2 | 175 | AAF18564.1 | |
| AW964442.1 | 3 | 186 | ||
| BC018969.1 | 2 | 146 | AAH18969.1 | |
| BC028388.1 | 3 | 208 | AAH28388.1 | |
| BF223532.1 | 3 | 159 | ||
| BG706927.1 | 2 | 230 | ||
| BI597156.1 | 3 | 226 | ||
| DB476616.1 | 3 | 264 |
More Information
Set Membership: |
|

Genomic Map