Assay Details
Target Gene Details
Entrez Gene ID: | 64783 |
Gene Name: | RNA binding motif protein 15 |
Gene Aliases: |
OTT, OTT1, SPEN |
Location: |
Chr.1:110338426-110373005 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM15 | NM_001201545.1 | 1 | 1955 | NP_001188474.1 |
| NM_022768.4 | 1 | 1955 | NP_073605.4 | |
| AB209680.1 | BAD92917.1 | |||
| AF368062.1 | 1 | 1955 | AAK54722.1 | |
| AF368063.1 | 1 | 1955 | AAK54723.1 | |
| AF368064.1 | 1 | 1955 | AAK54724.1 | |
| AK022541.1 | 1 | 1835 | BAB14088.1 | |
| AK025596.1 | 1 | 620 | BAB15185.1 | |
| BC006397.2 | 1 | 1812 | AAH06397.2 | |
| BC047479.1 | 1 | 1844 | AAH47479.1 | |
| BC062316.1 | 1 | 1812 | AAH62316.1 | |
| BC098140.1 | 1 | 1942 | AAH98140.1 | |
| BC103493.1 | 1 | 1942 | AAI03494.1 | |
| BC103507.1 | 1 | 1942 | AAI03508.1 |
More Information
Set Membership: |
|

Genomic Map