Assay Details
Target Gene Details
Entrez Gene ID: | 27069 |
Gene Name: | growth hormone inducible transmembrane protein |
Gene Aliases: |
DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5 |
Location: |
Chr.10:84139429-84153555 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GHITM | NM_014394.2 | 9 | 1715 | NP_055209.2 |
| AF078863.1 | 9 | 1639 | AAD44495.1 | |
| AK098770.1 | 10 | 1528 | ||
| AK130097.1 | 9 | 1583 | ||
| AK292864.1 | ||||
| AK298332.1 | ||||
| AL136713.1 | 9 | 1625 | CAB66648.1 | |
| AL834502.1 | 2 | 616 | CAH56398.1 | |
| AY358824.1 | 9 | 1590 | AAQ89183.1 | |
| BC010354.1 | 9 | 1586 | AAH10354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046749 | Chr.10:83270389 - 84629720 on Build GRCh38 | Gain |
|
| nsv551745 | Chr.10:83260366 - 84632635 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map