Assay Details
Target Gene Details
Entrez Gene ID: | 23597 |
Gene Name: | acyl-CoA thioesterase 9 |
Gene Aliases: |
ACATE2, CGI-16, MT-ACT48, MTACT48 |
Location: |
Chr.X:23701055-23743290 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT9 | NM_001033583.2 | NP_001028755.2 | ||
| NM_001037171.1 | NP_001032248.1 | |||
| XM_005274472.2 | XP_005274529.1 | |||
| AF132950.1 | AAD27725.1 | |||
| AF241787.1 | AAG44475.1 | |||
| AK000659.1 | ||||
| AK024337.1 | ||||
| BC136671.1 | ||||
| BC144360.1 | ||||
| BX648512.1 |
More Information
Set Membership: |
|

Genomic Map