Assay Details
Target Gene Details
Entrez Gene ID: | 11218 |
Gene Name: | DEAD-box helicase 20 |
Gene Aliases: |
DP103, GEMIN3 |
Location: |
Chr.1:111755568-111767577 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX20 | NM_007204.4 | 11 | 2931 | NP_009135.4 |
| AF106019.1 | 11 | 2584 | AAD42744.1 | |
| AF155803.1 | 11 | 2579 | AAK56799.1 | |
| AF171063.1 | 11 | 2867 | AAF14544.1 | |
| AK001506.1 | 11 | 2593 | BAA91727.1 | |
| AL133598.1 | 3 | 3755 | CAB63734.2 | |
| AY007146.1 | 1 | 534 | AAG02003.1 | |
| BC011556.1 | 11 | 2584 | AAH11556.1 | |
| BC031062.1 | 11 | 2593 | AAH31062.1 | |
| BC034953.2 | 11 | 2902 | AAH34953.1 |
More Information
Set Membership: |
|

Genomic Map