Assay Details
Target Gene Details
Entrez Gene ID: | 120939 |
Gene Name: | transmembrane protein 52B |
Gene Aliases: |
C12orf59 |
Location: |
Chr.12:10170599-10191804 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM52B | NM_001079815.1 | 6 | 741 | NP_001073283.1 |
| NM_153022.3 | 4 | 967 | NP_694567.1 | |
| XM_006719029.2 | 5 | 1011 | XP_006719092.1 | |
| AK055728.1 | 4 | 893 | BAB70998.1 | |
| AY358845.1 | 4 | 924 | AAQ89204.1 | |
| BC098571.1 | 5 | 451 | AAH98571.1 | |
| BC131523.1 | 5 | 455 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580102 | Chr.12:10186755 - 10190672 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map