Assay Details
Target Gene Details
Entrez Gene ID: | 101927550 |
Gene Name: | uncharacterized LOC101927550 |
Gene Aliases: |
- |
Location: |
Chr.7:99013165-99036492 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927550 | NR_110102.1 | |||
| AK001533.1 |
Target Gene Details
Entrez Gene ID: | 57154 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 1 |
Gene Aliases: |
- |
Location: |
Chr.7:99027435-99144120 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMURF1 | NM_001199847.1 | 18 | 4849 | NP_001186776.1 |
| NM_020429.2 | 19 | 4936 | NP_065162.1 | |
| NM_181349.2 | 18 | 4858 | NP_851994.1 | |
| XM_017012457.1 | 19 | 4925 | XP_016867946.1 | |
| AB046845.1 | 19 | 4904 | ||
| AK057024.1 | 1 | 1809 | ||
| AK126471.1 | 1 | 2802 | ||
| AL834242.1 | 7 | 3210 | CAD38919.1 | |
| AY007164.1 | 1 | 834 | ||
| BC152468.1 | 19 | 4904 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614267 | Chr.7:99027640 - 99036461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map