Assay Details
Target Gene Details
Entrez Gene ID: | 9 |
Gene Name: | N-acetyltransferase 1 |
Gene Aliases: |
AAC1, MNAT, NAT-1, NATI |
Location: |
Chr.8:18170419-18223689 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT1 | NM_000662.7 | 3 | 472 | NP_000653.3 |
| NM_001160170.3 | 5 | 1006 | NP_001153642.1 | |
| NM_001160171.3 | 4 | 856 | NP_001153643.1 | |
| NM_001160172.3 | 5 | 777 | NP_001153644.1 | |
| NM_001160173.3 | 4 | 622 | NP_001153645.1 | |
| NM_001160174.2 | 1 | 689 | NP_001153646.1 | |
| NM_001160175.3 | 5 | 740 | NP_001153647.1 | |
| NM_001160176.3 | 4 | 590 | NP_001153648.1 | |
| NM_001160179.2 | 5 | 676 | NP_001153651.1 | |
| NM_001291962.1 | 6 | 794 | NP_001278891.1 | |
| XM_006716410.3 | 3 | 1049 | XP_006716473.1 | |
| XM_011544687.1 | 6 | 1135 | XP_011542989.1 | |
| XM_011544688.1 | 5 | 985 | XP_011542990.1 | |
| XM_011544689.2 | 2 | 834 | XP_011542991.1 | |
| XM_017013947.1 | 8 | 1371 | XP_016869436.1 | |
| AJ581135.1 | CAE46162.1 | |||
| AJ581136.1 | CAE46163.1 | |||
| AJ581137.1 | CAE46164.1 | |||
| AJ581138.1 | CAE46165.1 | |||
| AJ581139.1 | CAE46166.1 | |||
| AJ581140.1 | CAE46167.1 | |||
| AJ581141.1 | CAE46168.1 | |||
| AK290912.1 | 3 | 947 | ||
| AV754344.1 | 3 | 469 | ||
| BC047666.1 | 3 | 451 | AAH47666.1 | |
| BG655073.1 | ||||
| BM924372.1 | 5 | 751 | ||
| BX647521.1 | 4 | 565 | ||
| D90041.1 | 1 | 455 | BAA14095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616514 | Chr.8:18121821 - 18287748 on Build GRCh38 | Gain |
|
| nsv1025858 | Chr.8:18177517 - 18279674 on Build GRCh38 | Loss |
|
| nsv428196 | Chr.8:18219826 - 18542377 on Build GRCh38 | Gain |
|
| nsv522647 | Chr.8:17979580 - 18314502 on Build GRCh38 | Loss |
|
| nsv831251 | Chr.8:18216369 - 18263220 on Build GRCh38 | Loss |
|
| nsv1026366 | Chr.8:18071966 - 18557180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map