Assay Details
Target Gene Details
Entrez Gene ID: | 51199 |
Gene Name: | ninein |
Gene Aliases: |
SCKL7 |
Location: |
Chr.14:50719763-50831121 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIN | NM_020921.3 | 31 | 6529 | NP_065972.3 |
| XM_011536817.1 | 31 | 6503 | XP_011535119.1 | |
| XM_011536822.1 | 30 | 4364 | XP_011535124.1 | |
| AB046785.2 | 31 | 4373 | BAB13391.2 | |
| AF186776.1 | 12 | 1658 | AAG17027.1 | |
| AF223938.1 | AAK00629.1 | |||
| AK025652.1 | 12 | 1649 | ||
| AK027054.1 | 4 | 559 | BAB15640.1 | |
| AY027794.1 | 12 | 1658 | AAK27375.1 | |
| AY027795.1 | 12 | 1658 | AAK27376.1 | |
| BM721510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564840 | Chr.14:50704161 - 50733474 on Build GRCh38 | Gain |
|
| nsv1040775 | Chr.14:50676760 - 50853094 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map