Assay Details
Target Gene Details
Entrez Gene ID: | 100289178 |
Gene Name: | GNG12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:67832288-68202987 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG12-AS1 | NR_040077.1 | |||
| AK124028.1 | BAC85755.1 |
Target Gene Details
Entrez Gene ID: | 79971 |
Gene Name: | wntless Wnt ligand secretion mediator |
Gene Aliases: |
C1orf139, EVI, GPR177, MRP, mig-14 |
Location: |
Chr.1:68098459-68233000 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WLS | NM_001002292.3 | NP_001002292.3 | ||
| NM_001193334.1 | NP_001180263.1 | |||
| NM_024911.6 | NP_079187.3 | |||
| XM_011542191.2 | XP_011540493.1 | |||
| XM_011542192.2 | XP_011540494.1 | |||
| XM_017002390.1 | XP_016857879.1 | |||
| AK026744.1 | BAB15540.1 | |||
| AK074583.1 | BAC11072.1 | |||
| AK074984.1 | BAC11333.1 | |||
| AK301613.1 | ||||
| AK309779.1 | ||||
| AY359035.1 | AAQ89394.1 | |||
| BC007211.1 | AAH07211.2 | |||
| BC110826.1 | AAI10827.1 | |||
| BC137109.1 | ||||
| BC137113.1 | ||||
| BX538320.1 | CAD98094.1 | |||
| BX648748.1 | ||||
| DQ323735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546448 | Chr.1:68072585 - 68177408 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map